Hydroxyurea, 98%
Hydroxyurea, 98%
Hydroxyurea, 98%
Thermo Scientific Chemicals

Hydroxyurea, 98%

Hydroxyurea, 98%, CAS # 127-07-1, is a compound with antineoplastic, immunomodulatory, radical scavenger, and antimitotic activities. It shows inhibitory activity against nucleic acid synthesis. | CAS: 127-07-1 | CH4N2O2 | 76.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 151680250
Price (USD)
388.65
Special offer
Online exclusive
Ends: 28-Dec-2026
457.00
Save 68.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
388.65
Special offer
Online exclusive
Ends: 28-Dec-2026
457.00
Save 68.35
Each
Chemical Identifiers
CAS127-07-1
IUPAC Namehydroxyurea
Molecular FormulaCH4N2O2
InChI KeyVSNHCAURESNICA-UHFFFAOYSA-N
SMILESNC(=O)NO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point140°C to 145°C
Titration Iodometric97.5 to 102.5 %
Loss on drying=<1 % (1 g, 105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Hydroxyurea is a member of the class of urea that shows antineoplastic, immunomodulatory, radical scavenger, and antimitotic activities.

• It has inhibitory activity against nucleic acid synthesis.

• It also inhibits ribonucleoside diphosphate reductase, an enzyme necessary to convert ribonucleoside diphosphate into deoxyribonucleoside diphosphate, preventing cells from leaving the G1/S phase of the cell cycle

Applications

• Hydroxyurea has an anti-retroviral activity, inhibiting human immunodeficiency virus type 1 (HIV-1) DNA synthesis in peripheral blood lymphocytes, decreasing the amount of intracellular deoxynucleotides

• This compound also has a radio sensitizing activity, maintaining cells in the radiation-sensitive G1 cell cycle phase and, consequently, interfering with DNA repair

• It has been used for cell cycle synchronization in human breast carcinoma cells

• This compound presents inhibitory activity in cell adhesion and motility assay in ccRCC Caki (HTB-47) and CD9 knockdown cells

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Sheehan, V. A.; Nelson, S.; Yappan, C.; Dinu, B. R.; Guffey, D.; Minard, C. G.; Ware, R. E. Hydroxyurea Improves Oxygen Transport Effectiveness in Sickle Cell Anemia Patients. Blood 2014, 121 (24), 2716-2716.
  2. Martinez-Rojano, H.; Mancilla-Ramirez, J.; Quiñonez-Diaz, L.; Galindo-Sevilla, N. Activity of hydroxyurea against Leishmania mexicana. Antimicrob Agents Chemother. 2008, 52, (10), 3642-3647.
  3. Garner, J.M.; Herr, M.J.; Hodges, K.B.; Jennings, L.K. The utility of tetraspanin CD9 as a biomarker for metastatic clear cell renal cell carcinoma. Biochem Biophys Res Commun. 2016, 471, (1), 21-25.
  4. Anele, U. A.; Mack, A. K.; Resar, L. M.; Burnett, A. L. Hydroxyurea therapy for priapism prevention and erectile function recovery in sickle cell disease: a case report and review of the literature. Int Urol Nephrol 2014, 46 (9), 1733-1736.