Hexanal, 96%
Hexanal, 96%
Hexanal, 96%
Thermo Scientific Chemicals

Hexanal, 96%

CAS: 66-25-1 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 151850025
Price (USD)
374.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
374.00
Each
Chemical Identifiers
CAS66-25-1
IUPAC Namehexanal
Molecular FormulaC6H12O
InChI KeyJARKCYVAAOWBJS-UHFFFAOYSA-N
SMILESCCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4020 to 1.4070 (20°C, 589 nm)
Density(20°C) 0.810 to 0.820 g/ml
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only