Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes
Thermo Scientific™

Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes

Enhance cell attachment to the culture surface with the extracellular matrix coating on Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsSurface Treatment
1520346Collagen Type I
1520356Poly-D-Lysine
15202524Poly-D-Lysine
Catalog number 152034
Price (CLP)
356.469
Case of 20
No. of Wells:
6
Surface Treatment:
Collagen Type I
Price (CLP)
356.469
Case of 20
Enhance cell attachment to the culture surface with the extracellular matrix coating on Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes, ideal for culturing fastidious cells that do not attach to the regular cell culture-treated surfaces.
Product overview
  • Aseptically manufactured
  • Poly-D-Lysine is a chemically synthesized animal-free material
  • Collagen Type I is from rat tail tendon
Poly-D-Lysine
Poly-D-Lysine (PDL) is a chemically synthesized extracellular matrix that is used to promote cell adhesion by altering surface charges on tissue-culture treated glass or plastic surfaces.  It specifically facilitates neuronal differentiation and expansion. PDL is animal origin-free and contains polymers with a molecular weight of 50,000–150,000 Daltons.

PDL is available in a coated multidish ready to use format to reduce preparation time. The plates can be stored at room temperature for a maximum shelf life of 24 months. PDL-coated plates are available as a case of 20 plates with 5 plates per vacuum sealed foil pack with a desiccant. Coating is done in a highly controlled, aseptic manufacturing environment that ensures lot-to-lot consistency and reproducibility.  Each lot is tested to help ensure we meet these standards.

Precoated plates save time & labor costs while increasing productivity and allowing you to focus on performing experiments. To meet all your application needs, PDL coating comes in a wide variety of formats - 6 well plate, 24 well plate, 96-well clear plate, 96-well black-walled imaging plate, 96-well white-walled imaging plate, 384-well black-walled imaging plate, T25, T75 and T175 flasks. PDL is used to promote adhesion of a variety of cell types like primary neurons, glial cells, PSC-derived neurons and transfected cells.

Collagen I
Collagen is the most abundant protein in mammals and a key structural component of several tissues. Collagen type I is the most commonly used extracellular matrix (ECM) protein for cell culture, facilitating cell attachment, growth, differentiation, migration, and tissue morphogenesis.

Collagen I, Rat Tail, is available in a coated multidish ready to use format with no preparation time. The plates are coated with Gibco™ Collagen I and can be stored at room temperature for a maximum shelf life of 12 months. Collagen I plates are available as a case of 20 plates with 5 plates per vacuum sealed foil pack with a desiccant. Coating is done in a highly controlled, aseptic manufacturing environment that ensures lot-to-lot consistency and reproducibility.  Each lot is tested to help ensure we meet these standards.

Precoated plates save time & labor costs while increasing productivity and allowing you to focus on performing experiments. To meet all your application needs, Collagen I coating comes in a wide variety of formats - 6 well plate, 96-well clear plate, 96 well black-walled imaging plate, 96-well white-walled imaging plate, T25, T75 and T175 flasks. Collagen I coated products are commonly used to culture endothelial cells, hepatocytes (human and rat), muscle cells, and a variety of other cell types.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Length (Metric)128 mm
Quantity5/Pk., 20/Cs.
Surface TreatmentCollagen Type I
Well ShapeFlat Bottom (F-well)
Width (Metric)86 mm
Culture Area9.6 sq. cm
No. of Wells6
SterilityAseptically Manufactured
TypeMulti Dish
Volume (Metric) Well3 mL (Well)
Volume (Metric) Working3 mL
Unit SizeCase of 20

Frequently asked questions (FAQs)

What does it mean for a plate to be "Aseptically Manufactured"?

Prior to treatment with an extra cellular matrix (ECM), the plates are sterilized. The ECM treatment is a process that utilizes sterile technique, but we cannot claim the product is "sterile" because it has been opened. If the product were to be re-sterilized after the treatment, the ECM coating would be ruined. For the purpose of using this plate in cell-culture applications, it is functionally sterile.