6 Well Plate, Poly-D-Lysine Coated Surface, Pack of 5 - FAQs
6 Well Plate, Poly-D-Lysine Coated Surface, Pack of 5 - FAQs
View additional product information for Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes - FAQs (152025, 152035, 152034)
1 product FAQs found
What does it mean for a plate to be "Aseptically Manufactured"?
Prior to treatment with an extra cellular matrix (ECM), the plates are sterilized. The ECM treatment is a process that utilizes sterile technique, but we cannot claim the product is "sterile" because it has been opened. If the product were to be re-sterilized after the treatment, the ECM coating would be ruined. For the purpose of using this plate in cell-culture applications, it is functionally sterile.