How many plates are contained in 96 Well Plate, Collagen I Coated Surface, Pack of 5 (Cat. No. 152038)?
96 Well Plate, Collagen I Coated Surface, Pack of 5 (Cat. No. 152038) provides 5 Cases with each Case containing 20 individual Nunc MicroWell Collagen Type I-coated 96-well microplates.
What does it mean for a plate to be "Aseptically Manufactured"?
Prior to treatment with an extra cellular matrix (ECM), the plates are sterilized. The ECM treatment is a process that utilizes sterile technique, but we cannot claim the product is "sterile" because it has been opened. If the product were to be re-sterilized after the treatment, the ECM coating would be ruined. For the purpose of using this plate in cell-culture applications, it is functionally sterile.