Triethyl 4-phosphonocrotonate, 92%, predominantly trans
Triethyl 4-phosphonocrotonate, 92%, predominantly trans
Triethyl 4-phosphonocrotonate, 92%, predominantly trans
Thermo Scientific Chemicals

Triethyl 4-phosphonocrotonate, 92%, predominantly trans

CAS: 10236-14-3 | C10H19O5P | 250.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 152120500
Price (USD)
301.65
Special offer
Online exclusive
Ends: 28-Dec-2026
355.00
Save 53.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
301.65
Special offer
Online exclusive
Ends: 28-Dec-2026
355.00
Save 53.35
Each
Chemical Identifiers
CAS10236-14-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=90 %
Appearance (Color)Clear light yellow
Infrared spectrumConforms
Refractive index1.4525 to 1.4555 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only