Search
View additional product information for Bgl II - FAQs (15213010)
4 product FAQs found
Enzyme - Units
Acc I - 4
Afl III - 0.5
Alu I - 1
AlwN I - 0.5
Ava I - 1
Ava II - 0.5 (a)
BstY I - 8
BamH I - 2
Ban II - 2
Bgl I - 8
Bsm I - 8
Cfo I - 4
Eco47 III - 8
Cla I - 1 (b)
Dde I - 1
Dra I - 4
EcoO109 I - 8
EcoR I - 0.5
EcoR II - >8 (a)
EcoRV - 2
Fsp I - 1
Hae II - 2
Hae III - 2
Hha I - 2
Hinc II - 4
Hind III - 0.5
Hinf I - 2
Hpa II - 1
Kpn2 I - 4
Mbo I - >8 (b)
Mbo II - 4 (b)
Msc I - >8 (a)
Mse I - 2
Msp I - 0.5
Nar I - 2
Nci I - 4
Nde I - 4
Nde II - >8 (b)
NgoA IV - 4
Nhe I - 2
Nsp I - 2
Nru I - 2 (b)
Psp5 II - 8
Pst I - 4
Pvu I - 2
Pvu II - 2
Rca I - 4
Rsa I - 4
Sal I - 8
Sau3A I - 2
Sau96 I - 8 (a)
Sca I - 4
Sph I - 2
Ssp I - 4
Sty I - 8
Taq I - 2 (b)
Tha I - 4
Vsp I - 0.5
Xma III - >8
(a) Sensitive to dcm methylation
(b) Sensitive to dam methylation
Notes:
- This list includes only restriction endonucleases with cleavage sites in pBR322, and not all enzymes listed are currently available for purchase from Thermo Fisher Scientific.
- In the experiments that generated this data, plasmid pBR322 was grown in E. coli RR1 and thus contained 5-methylcytosine and N6-methyladenine. Reactions contained 0.5, 1.0, 2.0, 4.0, or 8.0 units of enzyme, 1 µg of pBR322 DNA and the appropriate REACT Buffer in 20 µl. After a 1-h reaction, products were analyzed by electrophoresis on a 1% agarose gel.