Distilled Water Bag(s), 20 L - FAQs
Distilled Water Bag(s), 20 L - FAQs
View additional product information for Distilled Water - FAQs (15230279, 15230261, 15230253)
1 product FAQs found
What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?
These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.