Distilled Water, 500 mL - FAQs
Distilled Water, 500 mL - FAQs
View additional product information for Distilled Water - FAQs (15230089)
1 product FAQs found
Can I get the conductivity measurement for Distilled Water?
We do not test the “finished” product for conductivity as part of the QC testing specifications. However, conductivity testing is done in our water system on a daily basis. This is the water system used in the manufacture of cell culture media and reagents made at the Gibco site.
Conductivity testing is performed according to USP specifications. The USP specification is that conductivity be “not more than 1 micromho/cm”.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.