Distilled Water
Gibco™

Distilled Water

High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1000 mL
10 x 500 mL
20 x 100 mL
6 x 1 L
Catalog number 15230162
Price (USD)
21.65
Online exclusive
23.91
Save 2.26
Each
Quantity:
500 mL
Customize this product
Price (USD)
21.65
Online exclusive
23.91
Save 2.26
Each

This distilled water is high-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, this distilled water features:

  • Cell culture quality
  • Proven performance
  • Suitable for use in cell culture and molecular biology applications

Cell culture quality

This water is prepared by distillation and filtration (0.1 μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly.

Proven performance

The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.

Use in cell culture and molecular biology applications

This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers.

Dual-site cGMP manufacturing and quality system

This water is produced at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical product made in our Grand Island facility (15230-162). This facility is also registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
CAS7732-18-5
Chemical Name or MaterialDihydrogen monoxide
PackagingBottle
Purification MethodMembrane-Filtered
Recommended StorageStorage conditions: 15 – 30°C, Shipping conditions: Ambient, Shelf life: 24 months from date of manufacture
Shelf Life24 Months
Shipping ConditionRoom Temperature
SterilitySterile-filtered
Treatment(s)Not DEPC-Treated
GradeCell Culture
Molecular Weight (g/mol)18.02 g/mol
Quantity500 mL
Unit SizeEach

Frequently asked questions (FAQs)

Is Distilled Water (Cat. No. 15230147) RNAse-free?

Distilled Water (Cat. No. 15230147) is not RNAse-free. Instead, we would recommend using UltraPure DNase/RNase-Free Distilled Water (Cat. No. 10977049) to have it DNase/RNase-free.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I get the conductivity measurement for Distilled Water?

We do not test the “finished” product for conductivity as part of the QC testing specifications. However, conductivity testing is done in our water system on a daily basis. This is the water system used in the manufacture of cell culture media and reagents made at the Gibco site.

Conductivity testing is performed according to USP specifications. The USP specification is that conductivity be “not more than 1 micromho/cm”.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?

These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.