Distilled Water Bag(s), 5 L - FAQs
Distilled Water Bag(s), 5 L - FAQs
View additional product information for Distilled Water - FAQs (15230279, 15230261, 15230253)
7 product FAQs found
Can I get the conductivity measurement for Distilled Water?
We do not test the “finished” product for conductivity as part of the QC testing specifications. However, conductivity testing is done in our water system on a daily basis. This is the water system used in the manufacture of cell culture media and reagents made at the Gibco site.
Conductivity testing is performed according to USP specifications. The USP specification is that conductivity be “not more than 1 micromho/cm”.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?
These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will your company continue to support other films?
Yes. Although we feel that the PE film exceeds many of the technical properties of the other available films, we will continue to offer and support our other currently available films through custom orders.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
In what size package will my Gibco Cell Culture Bags ship?
Each bag size will arrive in a specific heavyweight corrugated box designed for shipment:
5 L: 12” x 14.25” x 5.5”
10 L: 13” x 23.5” x 6.125”
20 L: 15” x 24” x 6.125”
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the benefits of the Gibco PE film?
The commercially available film is a very clear, multi-layered, single web film with a polyethylene contact surface that can be made into a wide range of bag sizes (250 mL to 1,000L). In addition, the film is a market leader with exceptional gas barrier properties and very low extractable and leachable results. Click here to read more about the PE Film specifications (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/classical-media/gibco-cell-culture-bags.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the Gibco PE Film?
Based on feedback from customers, we designed a media bag portfolio that uses a consistent film for all bag sizes. This film, called PE film, is superior in function to other commercially available films. Please go here to read more about the PE Film specifications (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/classical-media/gibco-cell-culture-bags.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are Gibco Cell Culture Bags?
Gibco Cell Culture Bags are convenient, pre-filled bags in a variety of formulations and sizes, specifically designed to increase productivity and reduce costs. The bags can be formatted up to 1,000 L. Read more about these bags here (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/classical-media/gibco-cell-culture-bags.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.