2-Methyl-1,3-cyclopentanedione, 98%
2-Methyl-1,3-cyclopentanedione, 98%
2-Methyl-1,3-cyclopentanedione, 98%
Thermo Scientific Chemicals

2-Methyl-1,3-cyclopentanedione, 98%

CAS: 765-69-5 | C6H8O2 | 112.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 152321000
Price (USD)
296.65
Special offer
Online exclusive
Ends: 28-Dec-2026
349.00
Save 52.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
296.65
Special offer
Online exclusive
Ends: 28-Dec-2026
349.00
Save 52.35
Each
Chemical Identifiers
CAS765-69-5
IUPAC Name2-methylcyclopentane-1,3-dione
Molecular FormulaC6H8O2
InChI KeyHXZILEQYFQYQCE-UHFFFAOYSA-N
SMILESCC1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder
Melting point212°C to 216°C
Appearance (Color)Light beige to tan
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only