Antibiotic-Antimycotic (100X), 20 x 100 mL - FAQs
Antibiotic-Antimycotic (100X), 20 x 100 mL - FAQs
View additional product information for Antibiotic-Antimycotic (100X) - FAQs (15240112, 15240096, 15240062)
7 product FAQs found
Can I use antibiotics such as Pen-Strep or Antibiotic-Antimycotic (100X) when culturing Sf9 insect cells?
Yes, however, we do not recommend adding antibiotics to the medium at the time of thawing. Thaw cells into medium without antibiotic and allow the cells to recover from the thawing process. After that, you can add antibiotics by diluting into the medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the solvent used in Antibiotic-Antimycotic (100X)?
The components are in a 0.85% saline solution.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the components of Antibiotic-Antimycotic (100X)?
This solution contains 10,000 units/mL of penicillin, 10,000 µg/mL of streptomycin, and 25 µg/mL of Gibco Amphotericin B.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the recommended storage condition for Antibiotic-Antimycotic (100X)?
The recommended condition is storage at -5 to -20 degrees C and when stored as recommended, the shelf life is 12 months from the date of shipment.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Should I directly add 5 mL of Antibiotic-Antimycotic (100X) to 500 mL cell culture medium or should I dilute the Antibiotic-Antimycotic (100X) 100 times and then add 5 ml of the diluted Antibiotic-Antimycotic to 500 ml of cell culture medium?
The accurate way would be to dilute 5 mL of Antibiotic-Antimycotic (100X) into 495 mL of medium. Please note that the volume of medium in a 500 mL medium bottle is not exactly 500 mL. It could range from 500.5 - 510.5 mL.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How can I decontaminate my cultures?
When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.
1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.
The following is a suggested procedure for determining toxicity levels and decontaminating cultures:
1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What antibiotics do you offer to help control or eliminate cell culture contamination?
Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.