trans-Anethole, 99%
trans-Anethole, 99%
trans-Anethole, 99%
Thermo Scientific Chemicals

trans-Anethole, 99%

CAS: 4180-23-8 | C10H12O | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 152530050
Price (USD)
18.65
Special offer
Online exclusive
Ends: 28-Dec-2026
21.70
Save 3.05
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
18.65
Special offer
Online exclusive
Ends: 28-Dec-2026
21.70
Save 3.05
Each
Chemical Identifiers
CAS4180-23-8
IUPAC Name1-methoxy-4-[(1E)-prop-1-en-1-yl]benzene
Molecular FormulaC10H12O
InChI KeyRUVINXPYWBROJD-ONEGZZNKSA-N
SMILESCOC1=CC=C(\C=C\C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5580 to 1.5650 (20°C, 589 nm)
GC>=98.5 %
Density(25/4°C) 0.980 to 0.985
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only