Amaranth, 85%, pure
Amaranth, 85%, pure
Amaranth, 85%, pure
Thermo Scientific Chemicals

Amaranth, 85%, pure

CAS: 915-67-3 | C20H11N2Na3O10S3 | 604.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 153030250
Price (USD)
42.30
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
42.30
Each
Chemical Identifiers
CAS915-67-3
IUPAC Nametrisodium (4E)-3-oxo-4-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC20H11N2Na3O10S3
InChI KeyVOBHRQFELWTZFS-DXVNCNPQSA-K
SMILES[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C(=O)\C2=N\NC1=C2C=CC=CC2=C(C=C1)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red-brown to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
UV spectrophotometry>=80 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only