3,5-Dinitrobenzoic acid, 98+%
3,5-Dinitrobenzoic acid, 98+%
3,5-Dinitrobenzoic acid, 98+%
Thermo Scientific Chemicals

3,5-Dinitrobenzoic acid, 98+%

CAS: 99-34-3 | C7H4N2O6 | 212.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 153190010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS99-34-3
IUPAC Name3,5-dinitrobenzoic acid
Molecular FormulaC7H4N2O6
InChI KeyVYWYYJYRVSBHJQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point204°C to 209°C
Titration with NaOH98.0 to 102.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only