Giemsa Stain, pure, high purity biological stain
Giemsa Stain, pure, high purity biological stain
Giemsa Stain, pure, high purity biological stain
Thermo Scientific Chemicals

Giemsa Stain, pure, high purity biological stain

CAS: 51811-82-6 | C14H14ClN3S | 291.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 153230250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS51811-82-6
IUPAC Name7-amino-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
Molecular FormulaC14H14ClN3S
InChI KeyNALREUIWICQLPS-UHFFFAOYSA-N
SMILES[Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Specific absorbance A (1%/1cm)>=600 (in MeOH) On dried substance
Appearance (Color)Dark green to black
Infrared spectrumConforms
Loss on drying=<10 % (110°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only