Is the Oretronic IV Tramp Metal Detector (TMD) system more immune to false trips than the Oretronic III TMD system?
Yes, with factory and field testing, we have proven that the Oretronic IV TMD system is less affected by metal near the system and electrical interference by magnets, motors, and other equipment in the area, minimizing the occurrence of false trips.
Why is the Oretronic IV Tramp Metal Detector (TMD) system better than the Oretronic III TMD control?
The Oretronic IV Tramp Metal Detector (TMD) controls offer improved sensitivity, fewer false trips, better user interface, and multi-language software, compared to the Oretronic III TMD control.
Is the calibration process/menu the same for the Oretronic IV Tramp Metal Detector (TMD) controls as the previous Oretronic III TMD control?
The user interface and calibration menus have changed but the concept of operation and function of fine and coarse sensitivity remains the same.
Do the Oretronic IV Tramp Metal Detector (TMD) controls have the same mounting footprint as the previous Oretronic III TMD control?
No, the mounting footprint for the Oretronic IV TMD control versions is not the same as the Oretronic III TMD control. Please refer to the user manual (https://assets.thermofisher.com/TFS-Assets/CAD/manuals/tramp-metal-detector-oretronic-iv-s-manual-REC4479B.pdf) for dimensions and mounting requirements.
Is the Oretronic IV Tramp Metal Detector (TMD) control a direct replacement for the Oretronic III TMD control? Will it work with existing coils?
The Oretronic IV TMD control is a direct replacement for the Oretronic III TMD control and will work with existing Thermo Scientific transmitter, receiver, and under belt coils.