o-Toluoyl chloride, 99%
o-Toluoyl chloride, 99%
o-Toluoyl chloride, 99%
Thermo Scientific Chemicals

o-Toluoyl chloride, 99%

CAS: 933-88-0 | C8H7ClO | 154.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 153375000
Price (USD)
261.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
261.00
Each
Chemical Identifiers
CAS933-88-0
IUPAC Name2-methylbenzoyl chloride
Molecular FormulaC8H7ClO
InChI KeyGPZXFICWCMCQPF-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to pink
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5540 to 1.5560 (20°C, 589 nm)
Titration RCOCl>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only