D(+)-Malic acid, 98%
D(+)-Malic acid, 98%
D(+)-Malic acid, 98%
Thermo Scientific Chemicals

D(+)-Malic acid, 98%

CAS: 636-61-3 | C4H6O5 | 134.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 153500250
Price (USD)
318.65
Online Exclusive
354.00
Save 35.35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
318.65
Online Exclusive
354.00
Save 35.35 (10%)
Each
Chemical Identifiers
CAS636-61-3
IUPAC Name2-hydroxybutanedioic acid
Molecular FormulaC4H6O5
InChI KeyBJEPYKJPYRNKOW-UHFFFAOYNA-N
SMILESOC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Sulfated ash=<0.1 %
Specific optical rotation+1.8° to +2.6° (20°C, 589 nm) (c=3, Water)
HPLC=<1 % Fumaric acid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only