2-Amino-3-hydroxypyridine, 98%
2-Amino-3-hydroxypyridine, 98%
2-Amino-3-hydroxypyridine, 98%
Thermo Scientific Chemicals

2-Amino-3-hydroxypyridine, 98%

CAS: 16867-03-1 | C5H6N2O | 110.12 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 153570250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16867-03-1
IUPAC Name2-aminopyridin-3-ol
Molecular FormulaC5H6N2O
InChI KeyBMTSZVZQNMNPCT-UHFFFAOYSA-N
SMILESNC1=NC=CC=C1O
さらに表示
Appearance (Form)Powder or crystalline powder
Titration with HClO497.5 to 102.5 %
HPLC>=97.5 %
Appearance (Color)Beige to grey to brown
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only