Acetamide, 99%, pure
Acetamide, 99%, pure
Acetamide, 99%, pure
Thermo Scientific Chemicals

Acetamide, 99%, pure

CAS: 60-35-5 | C2H5NO | 59.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 153631000
Price (USD)
45.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
45.00
Each
Chemical Identifiers
CAS60-35-5
IUPAC Nameacetamide
Molecular FormulaC2H5NO
InChI KeyDLFVBJFMPXGRIB-UHFFFAOYSA-N
SMILESCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystals
Melting point78°C to 81°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only