Acetamide, 99%, pure
Acetamide, 99%, pure
Acetamide, 99%, pure
Thermo Scientific Chemicals

Acetamide, 99%, pure

CAS: 60-35-5 | C2H5NO | 59.06 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
產品號碼 153635000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS60-35-5
IUPAC Nameacetamide
Molecular FormulaC2H5NO
InChI KeyDLFVBJFMPXGRIB-UHFFFAOYSA-N
SMILESCC(N)=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystals
Melting point78°C to 81°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only