Allyl mercaptan, tech., 70%, remainder mainly sulfide, Thermo Scientific Chemicals
Allyl mercaptan, tech., 70%, remainder mainly sulfide, Thermo Scientific Chemicals
Allyl mercaptan, tech., 70%, remainder mainly sulfide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allyl mercaptan, tech., 70%, remainder mainly sulfide, Thermo Scientific Chemicals

CAS: 870-23-5 | C3H6S | 74.14 g/mol
Have Questions?
Catalog NumberQuantity
15371025025 g
Catalog number 153710250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS870-23-5
IUPAC Nameprop-2-ene-1-thiol
Molecular FormulaC3H6S
InChI KeyULIKDJVNUXNQHS-UHFFFAOYSA-N
SMILESSCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=65 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only