2-amino-4-picoline, 98%
2-amino-4-picoline, 98%
2-amino-4-picoline, 98%
Thermo Scientific Chemicals

2-amino-4-picoline, 98%

CAS: 695-34-1 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 153861000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS695-34-1
IUPAC Name4-methylpyridin-2-amine
Molecular FormulaC6H8N2
InChI KeyORLGLBZRQYOWNA-UHFFFAOYSA-N
SMILESCC1=CC=NC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Flakes or crystals or crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Water=<0.5 %
Appearance (Color)Light yellow to beige to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only