2-Heptanone, 98%
2-Heptanone, 98%
2-Heptanone, 98%
Thermo Scientific Chemicals

2-Heptanone, 98%

CAS: 110-43-0 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 154000010
Price (TWD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-43-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4075 to 1.4095 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only