cis-1,2-Cyclohexanedicarboxylic anhydride, 99%
cis-1,2-Cyclohexanedicarboxylic anhydride, 99%
cis-1,2-Cyclohexanedicarboxylic anhydride, 99%
Thermo Scientific Chemicals

cis-1,2-Cyclohexanedicarboxylic anhydride, 99%

CAS: 13149-00-3 | C8H10O3 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 154030025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13149-00-3
IUPAC Nameoctahydro-2-benzofuran-1,3-dione
Molecular FormulaC8H10O3
InChI KeyMUTGBJKUEZFXGO-UHFFFAOYNA-N
SMILESO=C1OC(=O)C2CCCCC12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Low melting fused solid
Infrared spectrumConforms
Melting point29°C to 34°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only