3-Bromochlorobenzene, 99%
3-Bromochlorobenzene, 99%
3-Bromochlorobenzene, 99%
Thermo Scientific Chemicals

3-Bromochlorobenzene, 99%

CAS: 108-37-2 | C6H4BrCl | 191.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 154200250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-37-2
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5760 to 1.5780 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only