4-Bromostyrene, 96%, stabilized
4-Bromostyrene, 96%, stabilized
4-Bromostyrene, 96%, stabilized
Thermo Scientific Chemicals

4-Bromostyrene, 96%, stabilized

CAS: 2039-82-9 | C8H7Br | 183.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
25 mL
100 mL
Packaging:
View all
Glass bottle
Catalog number 154280250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2039-82-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Liquid
Refractive index1.5920 to 1.5960 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only