3-tert-Butylphenol, 99+%
3-tert-Butylphenol, 99+%
3-tert-Butylphenol, 99+%
Thermo Scientific Chemicals

3-tert-Butylphenol, 99+%

CAS: 585-34-2 | C10H14O | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 154380250
Price (USD)
121.00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
121.00
Each
Chemical Identifiers
CAS585-34-2
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Crystalline powder
Melting point40°C to 43°C
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only