n-Butyrophenone, 99%
n-Butyrophenone, 99%
n-Butyrophenone, 99%
Thermo Scientific Chemicals

n-Butyrophenone, 99%

CAS: 495-40-9 | C10H12O | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 154411000
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Chemical Identifiers
CAS495-40-9
IUPAC Name1-phenylbutan-1-one
Molecular FormulaC10H12O
InChI KeyFFSAXUULYPJSKH-UHFFFAOYSA-N
SMILESCCCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5185 to 1.5205 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only