Nunc™ Lab-Tek™ II Chamber Slide™ System
Nunc™ Lab-Tek™ II Chamber Slide™ System
Nunc™ Lab-Tek™ II Chamber Slide™ System
Thermo Scientific™

Nunc™ Lab-Tek™ II Chamber Slide™ System

Grow cells directly on slides with the Nunc Lab-Tek II Chamber Slide System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWellsUnit Size
154461PK2Pack of 16
1544531Case of 96
154453PK
also known as 154453
1Pack of 16
1544612Case of 96
1545264Case of 96
154526PK4Pack of 16
1545348Case of 96
154534PK8Pack of 16
Catalog number 154461PK
Price (USD)
236.65
Online exclusive
247.00
Save 10.35
Pack of 16
Wells:
2
Unit Size:
Pack of 16
Price (USD)
236.65
Online exclusive
247.00
Save 10.35
Pack of 16

Grow, stain, and examine cells directly on the Thermo Scientific Nunc Lab-Tek II Chamber Slide, which consists of a removable polystyrene media chamber attached to a standard glass slide. The slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.

Features

  • Slide surface that is specially treated with a unique RS wash to ensure consistent cell growth
  • Tabbed polystyrene lid that allows gas exchange with minimal evaporation
  • Removal tool that separates media chamber from glass slide
  • Biocompatible adhesive hydrophobic well border printed on slide
  • White frosted writing area

Includes: chamber removal tool

Additional information

  • Can be purchased in case of 96 or pack of 16
  • Shipped sterile in convenient stackable incubation trays

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorClear
Description2-well Chamber Slide w/ removable wells
IncludesClear polystyrene lid with handles on the top and side for aseptic manipulations
Lid MaterialPolystyrene
No. per Pack16
Quantity1 Ea.
Culture Area4.0 cm2
Dimensions (LxW)74 x 25 mm
MaterialRS Glass
SterilitySterile
Surface TreatmentCell Culture
Volume (Metric) Working1.0 to 2.0 mL
Wells2
Unit SizePack of 16

Frequently asked questions (FAQs)

What cell density should I use for Lab-Tek Chambered Slides and Lab-Tek II Chambered Slides?

Unfortunately, we do not have a recommended cell density for our LabTek Chamber Slides. However, you can refer to the following link, which provides recommended cell densities for our other cell culture products:
https://www.thermofisher.com/us/en/home/references/gibco-cell-culture-basics/cell-culture-protocols/cell-culture-useful-numbers.html

Are your Nunc Lab-Tek II Chamber Slide System products pre-coated?

The Nunc Lab-Tek II Chamber Slide System products are not coated with any chemicals or reagents.

What is the difference between your original Nunc Lab-Tek Chamber Slide System and your Nunc Lab-Tek II Chamber Slide System?

Please review the following key differences between the Nunc Lab-Tek Chamber Slide System and the Nunc Lab-Tek II Chamber Slide System:
- The Nunc Lab-Tek Chamber Slide System is available in Permanox plastic or glass and incorporates a silicone gasket.
- The Nunc Lab-Tek II Chamber Slide System is available in glass only and do not have a silicone gasket.
- The Nunc Lab-Tek II Chamber Slide System has a blue hydrophobic border and the upper structure is attached by an adhesive. Supplied with this product is a key and holder system which is used to remove the upper structure from the glass slide.
- The Nunc Lab-Tek Chamber Slide System is available in 1-, 2-, 4-, 8- or 16-well formats.
- The Nunc Lab-Tek II Chamber Slide System is available in 1-, 2-, 4- or 8-well formats.

On the packaging of your Nunc Lab-Tek II Chamber Slide System, it states these chamber slides have an RS-treated growth surface. What is RS treatment?

The RS treatment is a wash step formulated to improve the consistency of the glass surface of the Nunc Lab-Tek II Chamber Slide System. RS glass treatment prepares the glass surface for culture after the hydrophobic border has been applied.
The term "RS treatment" refers to the glass washing procedure.

What stains and fixatives are compatible with Lab-Tek chambered slides and coverglasses?

Lab-Tek products are compatible with all common fixatives, including paraformaldehyde. Acetone at 100% and some mixtures of acetone and alcohols are not compatible with the polystyrene upper well structure and will result in clouding or other effects of chemical attack. We encourage careful testing within planned operating conditions before working with high-value samples.