2-Chlorobenzaldehyde, 99%
2-Chlorobenzaldehyde, 99%
2-Chlorobenzaldehyde, 99%
Thermo Scientific Chemicals

2-Chlorobenzaldehyde, 99%

CAS: 89-98-5 | C7H5ClO | 140.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
500 mL
1 L
Packaging:
Glass bottle
Catalog number 154611000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS89-98-5
IUPAC Name2-chlorobenzaldehyde
Molecular FormulaC7H5ClO
InChI KeyFPYUJUBAXZAQNL-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Free acid=<1 % (as chlorobenzoic acid)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only