Pierce™ Streptavidin Coated High Capacity Plates, Clear, 96-Well, 5 plates - FAQs
Pierce™ Streptavidin Coated High Capacity Plates, Clear, 96-Well, 5 plates - FAQs
View additional product information for Pierce™ Streptavidin Coated High Capacity Plates - FAQs (15502, 15501, 15500, 15506, 15505, 15504, 15503)
2 product FAQs found
How has Streptavidin been linked to the Pierce Streptavidin Coated High Capacity Plates (Cat. No. 15500)?
The linking of streptavidin has been performed by passive binding. This means the molecule has been attached to the plates just by simple incubation (see also https://www.thermofisher.com/de/en/home/life-science/protein-biology/protein-assays-analysis/elisa/elisa-microplates-plasticware/adsorption-immunoassay-plates.html).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the shelf life of Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well?
Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale for more details.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.