Pierce™ Streptavidin Coated High Capacity Plates
Pierce™ Streptavidin Coated High Capacity Plates
Pierce™ Streptavidin Coated High Capacity Plates
Thermo Scientific™

Pierce™ Streptavidin Coated High Capacity Plates

Thermo Scientific™ Pierce Streptavidin High Binding Capacity (HBC) Coated Plates are designed for binding biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of WellsFormatDetection Method
15504Clear384StandardColorimetric
15503Black96StandardFluorescence
15506Black384StandardFluorescence
15501Clear96Strip PlateColorimetric
15500Clear96StandardColorimetric
15502White96StandardChemiluminescence
15505White384StandardChemiluminescence
Catalog number 15504
Price (USD)
527.65
Online exclusive
614.00
Save 86.35
Each
Color:
Clear
No. of Wells:
384
Format:
Standard
Detection Method:
Colorimetric
Request bulk or custom format
Price (USD)
527.65
Online exclusive
614.00
Save 86.35
Each

Pierce Streptavidin High Binding Capacity (HBC) Plates are prepared with an exclusive coating method that yields a streptavidin-coated plate with four to five times the binding capacity of other commercially available polystyrene assay plates. Using a Pierce Streptavidin HBC Plate can result in an assay with a broader dynamic range and better linearity, leading to improved assay precision. The plates are offered in several plate formats, plate colors, and coating volumes.

Thermo Scientific Pierce Streptavidin-Coated High-Capacity Plates are designed for the binding of biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.

Pierce Streptavidin-coated plates are widely used for applications such as ELISAs and other assay protocols. These biotin-binding plates come with different capacities and sensitivities for maximum flexibility. For example, the high-binding-capacity plates (HBC) can be used for anti-therapeutic/anti-drug antibodies (ADA) screening for immunogenicity research and biologics development (1,2). Both regular and HBC plates have been used in aptamer assays for clinical and industrial needs, along with neurobiology research for therapeutics and prevention, amongst other applications (3–5).

Features of Streptavidin-Coated High-Capacity Plates include:

  • Unique plate-coating technology—results in high loading and binding capacity
  • Streptavidin-coated option—plates have up to five times the binding capacity of typical plates
  • Broader dynamic range—extends the quantitative range so there is less need for dilutions
  • Save time—pre-blocked plates to reduce the number of assay steps
  • Flexible assay formats—coated plates offered in 8-well strips to reduce waste

The proprietary plate-coating technology used to manufacture these Pierce coated plates yields high-capacity plates with a wider detection limit than regular-binding-capacity plates. The standard curve exhibits greater linearity for detecting small biotinylated molecules such as peptides and oligonucleotides, resulting in greater assay precision.

References

  1. J Immunol Methods. 2016 Apr;431:45-51
  2. J Immunol Methods. 2011 Feb 28;365(1-2):38-49
  3. Cancers (Basel). 2022 Oct 25;14(21):5222
  4. J Microbiol Biotechnol. 2013 Jun 28;23(6):878-84
  5. Acta Neuropathol Commun. 2018 May 31;6(1):43
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyHigh Capacity, Affinity
Detection MethodColorimetric
FormatStandard
No. per Pack5 Plates
Plate BlockingSuperBlock™ Buffer
Product TypeMicroplate
Quantity5 plates
ColorClear
MaterialPolystyrene
No. of Wells384
Product LinePierce
Surface TreatmentStreptavidin
Well ShapeFlat Bottom (F-well)
Unit SizeEach
Contents & Storage
Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C.

Frequently asked questions (FAQs)

How has Streptavidin been linked to the Pierce Streptavidin Coated High Capacity Plates (Cat. No. 15500)?

The linking of streptavidin has been performed by passive binding. This means the molecule has been attached to the plates just by simple incubation (see also https://www.thermofisher.com/de/en/home/life-science/protein-biology/protein-assays-analysis/elisa/elisa-microplates-plasticware/adsorption-immunoassay-plates.html).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is the shelf life of Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well?

Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale for more details.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.