Dibutyl phosphite, 14.5-16% P
Dibutyl phosphite, 14.5-16% P
Dibutyl phosphite, 14.5-16% P
Thermo Scientific Chemicals

Dibutyl phosphite, 14.5-16% P

CAS: 1809-19-4 | C8H19O3P | 194.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
155061000100 gGlass bottle
Catalog number 155061000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1809-19-4
IUPAC Namedibutoxy(oxo)-λ⁵-phosphanylium
Molecular FormulaC8H18O3P
InChI KeyOSPSWZSRKYCQPF-UHFFFAOYSA-N
SMILESCCCCO[P+](=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Assay14.5 to 16 % (P)
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4225 to 1.4255 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only