Pierce™ NeutrAvidin™ Coated High Capacity Plates
Thermo Scientific™

Pierce™ NeutrAvidin™ Coated High Capacity Plates

Thermo Scientific™ Pierce NeutrAvidin High Binding Capacity (HBC) Coated Plates are pre-blocked and ready-to-use polystyrene microplates for assays involving biotinylated antibodies or biotin-labeled molecules.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of WellsFormatDetection Method
15509White96StandardChemiluminescence
15510Black96StandardFluorescence
15513Black384StandardFluorescence
15508Clear96Strip PlateColorimetric
15507Clear96StandardColorimetric
Catalog number 15509
Price (USD)
287.65
Online exclusive
309.00
Save 21.35
Each
Color:
White
No. of Wells:
96
Format:
Standard
Detection Method:
Chemiluminescence
Request bulk or custom format
Price (USD)
287.65
Online exclusive
309.00
Save 21.35
Each
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. With a near-neutral pI (6.3), this results in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein lacks the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells.

Thermo Scientific NeutrAvidin™ Coated High Capacity Plates are designed for binding biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.

Features of Thermo Scientific Pierce Coated Plates include:

  • Unique plate-coating technology – results in high loading and binding capacity
  • NeutrAvidin Protein – coated plates have up to twice the binding capacity of typical plates
  • Robust and sensitive – NeutrAvidin Protein yields less nonspecific binding for improved signal-to-noise ratios
  • Broader dynamic range – extends the quantitative range so there is less need for dilutions
  • Save time – pre-blocked plates to reduce the number of assay steps
  • Flexible assay formats – choose from 96- and 384-well formats in a variety of colors

The proprietary plate-coating technology used to manufacture these Thermo Scientific Pierce Coated Plates yields high-capacity plates with a wider detection limit than regular binding capacity plates. The standard curve exhibits greater linearity for detecting small biotinylated molecules such as peptides and oligonucleotides, resulting in greater assay precision.

For detection of low concentrations (ng/mL) of biotinylated antibodies and other proteins, check out

Pierce™ NeutrAvidin™ Coated High Sensitivity Plates, Clear, 8-Well Strip

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyHigh Capacity, Affinity
Detection MethodChemiluminescence
FormatStandard
No. per Pack5 Plates
Plate BlockingSuperBlock™ Buffer
Product TypeMicroplate
Quantity5 plates
ColorWhite
MaterialPolystyrene
No. of Wells96
Product LineNeutrAvidin Pierce
Surface TreatmentNeutrAvidin Protein
Unit SizeEach
Contents & Storage
Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C.