UltraPure™ TEMED - FAQs
View additional product information for UltraPure™ TEMED - FAQs (15524010)
1 product FAQs found
Do you carry ammonium persulfate and TEMED?
Yes, details below:
- Ammonium Persulfate: Cat. No. 17874
- TEMED: Cat. No. 15524010 and Cat. No. 17919
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.