2,6-Dimethylmorpholine, 97%, mixture of isomers
2,6-Dimethylmorpholine, 97%, mixture of isomers
2,6-Dimethylmorpholine, 97%, mixture of isomers
Thermo Scientific Chemicals

2,6-Dimethylmorpholine, 97%, mixture of isomers

CAS: 141-91-3 | C6H13NO | 115.17 g/mol
Have Questions?
Catalog NumberQuantity
155341000100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 155341000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS141-91-3
IUPAC Name2,6-dimethylmorpholine
Molecular FormulaC6H13NO
InChI KeyHNVIQLPOGUDBSU-UHFFFAOYNA-N
SMILESCC1CNCC(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=96.0 % (mixture of isomers)
Refractive index1.4435 to 1.4485 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only