S.O.C. Medium - FAQs
S.O.C. Medium - FAQs
View additional product information for S.O.C. Medium - FAQs (15544034)
4 product FAQs found
How do you recommend that I prepare my DNA for successful electroporation of E. coli?
For best results, DNA used in electroporation must have a very low ionic strength and a high resistance. A high-salt DNA sample may be purified by either ethanol precipitation or dialysis.
The following suggested protocols are for ligation reactions of 20ul. The volumes may be adjusted to suit the amount being prepared.
Purifying DNA by Precipitation: Add 5 to 10 ug of tRNA to a 20ul ligation reaction. Adjust the solution to 2.5 M in ammonium acetate using a 7.5 M ammonium acetate stock solution. Mix well. Add two volumes of 100 % ethanol. Centrifuge at 12,000 x g for 15 min at 4C. Remove the supernatant with a micropipet. Wash the pellet with 60ul of 70% ethanol. Centrifuge at 12,000 x g for 15 min at room temperature. Remove the supernatant with a micropipet. Air dry the pellet. Resuspend the DNA in 0.5X TE buffer [5 mM Tris-HCl, 0.5 mM EDTA (pH 7.5)] to a concentration of 10 ng/ul of DNA. Use 1 ul per transformation of 20 ul of cell suspension.
Purifying DNA by Microdialysis: Float a Millipore filter, type VS 0.025 um, on a pool of 0.5X TE buffer (or 10% glycerol) in a small plastic container. Place 20ul of the DNA solution as a drop on top of the filter. Incubate at room temperature for several hours. Withdraw the DNA drop from the filter and place it in a polypropylene microcentrifuge tube. Use 1ul of this DNA for each electrotransformation reaction.
What is the formulation of the SOC medium that is provided with competent cells?
SOC (Super Optimal Catabolite) Medium Preparation (for 1 Liter):
1) To a 2 Liter flask with stir bar add the following:
- Bacto Tryptone 20 g
- Yeast Extract 5 g
- Sodium Chloride (NaCl) 0.58 g
- Potassium Chloride (KCl) 0.186 g
2) Add sterile water to a final volume of 1 Liter.
3) Mix well on magnetic stir plate for 5-10 minutes or until all of the ingredients are well mixed and completely dissolved.
4) Autoclave 30 minutes.
5) Allow to cool to room temperature.
6) Add 10 ml of sterile 2M Magnesium Solution (1M Magnesium sulfate, 1M Magnesium chloride)and mix well.
7) Add 10 ml of sterile 2M Glucose and mix well. (Final Glucose concentration is 20 mM).
Is S.O.C. medium absolutely required when recovering competent bacterial cells during transformation?
Many media can be used to grow transformed cells, including standard LB, SOB or TB broths. However, S.O.C. is the optimal choice for recovery of the cells before plating. The nutrient-rich formula with added glucose is often important for obtaining maximum transformation efficiencies.
When should DMSO, formamide, glycerol and other cosolvents be used in PCR?
Cosolvents may be used when there is a failure of amplification, either because the template contains stable hairpin-loops or the region of amplification is GC-rich. Keep in mind that all of these cosolvents have the effect of lowering enzyme activity, which will decrease amplification yield. For more information see P Landre et al (1995). The use of co-solvents to enhance amplification by the polymerase chain reaction. In: PCR Strategies, edited by MA Innis, DH Gelfand, JJ Sninsky. Academic Press, San Diego, CA, pp. 3-16.
Additionally, when amplifying very long PCR fragments (greater than 5 kb) the use of cosolvents is often recommended to help compensate for the increased melting temperature of these fragments.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.