1-Dodecanol, 98%
1-Dodecanol, 98%
1-Dodecanol, 98%
Thermo Scientific Chemicals

1-Dodecanol, 98%

CAS: 112-53-8 | C12H26O | 186.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
500 mL
1 L
10 L
Catalog number 155450010
Price (USD)
68.65
Special offer
Online exclusive
Ends: 28-Dec-2026
80.70
Save 12.05
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 28-Dec-2026
80.70
Save 12.05
Each
Chemical Identifiers
CAS112-53-8
IUPAC Namedodecan-1-ol
Molecular FormulaC12H26O
InChI KeyLQZZUXJYWNFBMV-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Liquid or crystalline solid
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only