Isoamylamine, 98+%
Isoamylamine, 98+%
Isoamylamine, 98+%
Thermo Scientific Chemicals

Isoamylamine, 98+%

CAS: 107-85-7 | C5H13N | 87.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 155470050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-85-7
IUPAC Name3-methylbutan-1-amine
Molecular FormulaC5H13N
InChI KeyBMFVGAAISNGQNM-UHFFFAOYSA-N
SMILESCC(C)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.5 %
Appearance (Form)Liquid
GC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only