How should I store UltraPure 1 M Tris-HCI Buffer, pH 7.5 (Cat. No. 15567027)?
We recommend storing UltraPure 1 M Tris-HCI Buffer, pH 7.5 (Cat. No. 15567027) in the refrigerator (2-8 degrees C).
Search
View additional product information for UltraPure™ 1 M Tris-HCI Buffer, pH 7.5 - FAQs (15567027)
2 product FAQs found