UltraPure™ 1M Tris-HCI, pH 8.0 - FAQs
UltraPure™ 1M Tris-HCI, pH 8.0 - FAQs
View additional product information for UltraPure™ 1M Tris-HCI, pH 8.0 - FAQs (15568025)
2 product FAQs found
How should I store UltraPure 1M Tris-HCI, pH 8.0 (Cat. No. 15568025)?
We recommend storing UltraPure 1M Tris-HCI, pH 8.0 (Cat. No. 15568025) in the refrigerator (2-8 degrees C).
What alternative do you offer for UltraPure 1M Tris-HCI, pH 8.0 (Cat. No. 15568025)?
We recommend Tris (1 M), pH 8.0, RNase‐free (Cat. No. AM9855G).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.