I received a shipment of UltraPure 0.5M EDTA, pH 8.0 (Cat. No. 15575020), but it was stored below 15-30 degrees C over the weekend. Can you confirm this product will still be stable?
We do not have specific data for temperatures below 15 degrees C for UltraPure 0.5M EDTA, pH 8.0 (Cat. No. 15575020). However, as long as the product was not frozen, the performance should not be affected.