Can I use EDTA (Cat. No. 17892) in molecular biology? If not, can you provide an alternative?
EDTA (Cat. No. 17892) is not intended for molecular biology applications.
UltraPure Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate (Na2EDTA·2H2O) (Cat. No. 15576028) is a potential alternative intended for molecular biology applications.
UltraPure Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate (Na2EDTA·2H2O) (Cat. No. 15576028) is a potential alternative intended for molecular biology applications.