3-Pentanone, 98%, pure
3-Pentanone, 98%, pure
3-Pentanone, 98%, pure
Thermo Scientific Chemicals

3-Pentanone, 98%, pure

CAS: 96-22-0 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 155810025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS96-22-0
IUPAC Namepentan-3-one
Molecular FormulaC5H10O
InChI KeyFDPIMTJIUBPUKL-UHFFFAOYSA-N
SMILESCCC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.3910 to 1.3930 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only