trans-Crotonyl chloride, 90%, tech.
trans-Crotonyl chloride, 90%, tech.
trans-Crotonyl chloride, 90%, tech.
Thermo Scientific Chemicals

trans-Crotonyl chloride, 90%, tech.

CAS: 625-35-4 | C4H5ClO | 104.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 155911000
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
141.00
Save 21.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
141.00
Save 21.35
Each
Chemical Identifiers
CAS625-35-4
IUPAC Name(2E)-but-2-enoyl chloride
Molecular FormulaC4H5ClO
InChI KeyRJUIDDKTATZJFE-NSCUHMNNSA-N
SMILESC\C=C\C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=88 %
Appearance (Color)Clear colorless to brown
Infrared spectrumConforms
Refractive index1.4585 to 1.4615 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only