4-Methylbenzyl alcohol, 98%
4-Methylbenzyl alcohol, 98%
4-Methylbenzyl alcohol, 98%
Thermo Scientific Chemicals

4-Methylbenzyl alcohol, 98%

CAS: 589-18-4 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 155920250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS589-18-4
IUPAC Name(4-methylphenyl)methanol
Molecular FormulaC8H10O
InChI KeyKMTDMTZBNYGUNX-UHFFFAOYSA-N
SMILESCC1=CC=C(CO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystals or needles or crystalline mass
Melting point56°C to 61°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only