Particle Certified HDPE Containers
Actual product may vary
Thermo Scientific™

Particle Certified HDPE Containers

Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Certified
156-125WNLP125 mLLow Particulate
156-125WNLPM
also known as 156-125W/N/LPM
125 mLLow Particulate, Low Metals
157-250WNLPM
also known as 157-250W/N/LPM
250 mLLow Particulate, Low Metals
157-250WNLP250 mLLow Particulate
150-01WNLPM
also known as 150-01W/N/LPM
1000 mLLow Particulate, Low Metals
150-01WNLP1000 mLLow Particulate
Catalog number 156-125WNLP
Price (USD)
322.00
Each
Capacity (Metric):
125 mL
Certified:
Low Particulate
Price (USD)
322.00
Each

Protect the quality of your products or lab samples with reliable Thermo Scientific™ Particle-Certified HDPE Containers, tested and certified to contain <20 particles per milliliter = 0.3 microns. A Certificate of Analysis is included with each shipment.

A large variety of standard particle-certified containers are available. Nonstandard containers may be custom cleaned on request.

Key Features:

  • Narrow-mouth
  • Includes leak-proof† polypropylene caps
  • Suitable for use in sampling and packaging, including combination packaging for shipping of hazardous materials
  • Double-bagged and ready for cleanroom with no extra preparation

Recommended for:

Low particle applications

†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity4 oz., 125 mL
Capacity (English)4 oz.
Certifications/ComplianceLow metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
CertifiedLow Particulate
Closure MaterialPolypropylene
Closure Size24-415 mm
Diameter (English)2 in.
Diameter (Metric)50.8 mm
Height (English)4 in.
Height (Metric)101.6 mm
No. per Case24
Product LineNalgene
Product TypeWater Sample Collection Bottle
ShapeRound
Volume (English) Brim4 oz.
Volume (Metric) Brim125 mL
Workflow StepUpstream process solution storage. Upstream Validation. Upstream process solution storage
Capacity (Metric)125 mL
Closure IncludedYes
ColorTranslucent
MaterialHDPE
MouthNarrow
Unit SizeEach